accessibility menu, dialog, popup

UserName

CH3ONHHNCH3OOO

CAS RN 205873-56-9

IUPAC Name: 3,4-diacetamidobenzoate
Synonyms: 3,4-diacetamidobenzoate
Molecular Weight (g/mol): 235.22
Molecular Formula: C11H11N2O4
InChi Key: GHTLCMPYQOFKCS-UHFFFAOYSA-M
SMILES: CC(=O)NC1=CC=C(C=C1NC(C)=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results