accessibility menu, dialog, popup

UserName

INCS

CAS RN 2059-76-9

IUPAC Name: 1-iodo-4-isothiocyanatobenzene
Synonyms: 1-iodo-4-isothiocyanatobenzene
Molecular Weight (g/mol): 261.08
Molecular Formula: C7H4INS
InChi Key: SNLGBRZZFRSXHA-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)N=C=S
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results