accessibility menu, dialog, popup

UserName

CAS RN 20595-45-3

HOOClCl(E)

CAS RN 20595-45-3

IUPAC Name: (2E)-3-(2,4-dichlorophenyl)prop-2-enoic acid
Synonyms: 2,4-dichlorocinnamic acid
Molecular Weight (g/mol): 217.05
Molecular Formula: C9H6Cl2O2
InChi Key: MEBWABJHRAYGFW-DUXPYHPUSA-N
SMILES: OC(=O)\C=C\C1=CC=C(Cl)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results