CAS RN 206658-83-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sodium 4-aminohippurate hydrate, 98%
CAS: 206658-83-5 Molecular Formula: C9H10N2NaO3+ Molecular Weight (g/mol): 217.18 MDL Number: MFCD00150723 InChI Key: UNZMYCAEMNVPHX-UHFFFAOYSA-N Synonym: aminohippurate sodium PubChem CID: 57465078 IUPAC Name: sodium;2-[(4-aminobenzoyl)amino]acetic acid SMILES: C1=CC(=CC=C1C(=O)NCC(=O)O)N.[Na+]