accessibility menu, dialog, popup

UserName

CAS RN 207386-92-3

NOHSOOHO

CAS RN 207386-92-3

IUPAC Name: 8-hydroxyquinoline-5-sulfonic acid hydrate
Synonyms: 8-hydroxyquinoline-5-sulfonic acid hydrate
Molecular Weight (g/mol): 243.23
Molecular Formula: C9H9NO5S
InChi Key: WUXYGIQVWKDVTJ-UHFFFAOYSA-N
SMILES: O.OC1=CC=C(C2=CC=CN=C12)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results