missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline-5-sulfonic acid hydrate, 98%

Product Code. 11354629
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354629 500 g 500g
11470481 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354629 Supplier Thermo Scientific Alfa Aesar Supplier No. B20022.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207386-92-3 | C9H7NO4S | 225.22 g/mol

It is employed as a reagent for determination of trace metals. It is employed as pre-1,2 and post-column3 reagent for fluorescence detection of metal ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reagent for determination of trace metals. It is employed as pre-1,2 and post-column3 reagent for fluorescence detection of metal ions.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207386-92-3
Molecular Formula C9H7NO4S
Molecular Weight (g/mol) 225.22
MDL Number MFCD00149482
InChI Key LGDFHDKSYGVKDC-UHFFFAOYSA-N
Synonym 8-hydroxyquinoline-5-sulfonic acid hydrate, 8-hydroxy-5-quinolinesulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid monohydrate, unii-n85kc14z44, 8-hydroxy-5-quinolinesulfonic acid hydrate, 5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1, 8-hydroxyquinoline-5-sulfonic acid, hydrate, acmc-209pxa, dsstox_cid_24317, dsstox_rid_80150
PubChem CID 2723649
IUPAC Name 8-hydroxyquinoline-5-sulfonic acid;hydrate
SMILES OC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O

Specifications

Melting Point 311°C to 313°C
Quantity 500 g
Beilstein 195247
Merck Index 14,4844
Solubility Information Soluble in water.
Formula Weight 225.22 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 8-Hydroxyquinoline-5-sulfonic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.