accessibility menu, dialog, popup

UserName

ClONS

CAS RN 20751-75-1

IUPAC Name: 2-chloro-1-(3,4-dihydro-2H-1,4-benzothiazin-4-yl)ethan-1-one
Synonyms: 2-chloro-1-(2,3-dihydro-1,4-benzothiazin-4-yl)ethanone
Molecular Weight (g/mol): 227.71
Molecular Formula: C10H10ClNOS
InChi Key: AYFZGNOUGDUMDS-UHFFFAOYSA-N
SMILES: ClCC(=O)N1CCSC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results