missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloroacetyl)-2H-1,4-benzothiazine, 97%, Thermo Scientific™

Product Code. 15477588
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477588 250 mg 250mg
15467588 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477588 Supplier Thermo Scientific Alfa Aesar Supplier No. H61095.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20751-75-1
Molecular Formula C10H10ClNOS
Molecular Weight (g/mol) 227.706
MDL Number MFCD06655687
InChI Key AYFZGNOUGDUMDS-UHFFFAOYSA-N
Synonym 4-chloroacetyl-3,4-dihydro-2h-1,4-benzothiazine, 4-2-chloroacetyl-2h-1,4-benzothiazine, 2-chloro-1-3,4-dihydro-2h-1,4-benzothiazin-4-yl ethan-1-one, 2-chloro-1-2,3-dihydro-1,4-benzothiazin-4-yl ethanone, ethanone, 2-chloro-1-2,3-dihydro-4h-1,4-benzothiazin-4-yl, 2-chloro-1-10-thia-7-azabicyclo 4.4.0 deca-1,3,5-trien-7-yl ethanone, 1-2h-benzo b 1,4 thiazin-4 3h-yl-2-chloroethanone, 2-chloro-1-2,3-dihydro-4h-1,4-benzothiazin-4-yl ethan-1-one
PubChem CID 4962259
IUPAC Name 2-chloro-1-(2,3-dihydro-1,4-benzothiazin-4-yl)ethanone
SMILES C1CSC2=CC=CC=C2N1C(=O)CCl

Specifications

Quantity 250 mg
Formula Weight 227.71
Percent Purity 97%
Chemical Name or Material 4-(2-Chloroacetyl)-2H-1,4-benzothiazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.