accessibility menu, dialog, popup

UserName

CAS RN 207605-40-1

CH3SOOO

CAS RN 207605-40-1

IUPAC Name: pentane-1-sulfonate
Synonyms: pentane-1-sulfonate
Molecular Weight (g/mol): 151.2
Molecular Formula: C5H11O3S
InChi Key: RJQRCOMHVBLQIH-UHFFFAOYSA-M
SMILES: CCCCCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)