missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic acid, sodium salt monohydrate, 98+%, HPLC grade

Product Code. 10485661 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10485661 100 g Glass bottle 100g
10178230 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10485661 Supplier Thermo Scientific Supplier No. 423201000
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 207605-40-1 | C5H11NaO3S·H2O | 192.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207605-40-1
Molecular Formula C5H11NaO3S·H2O
Molecular Weight (g/mol) 192.22
MDL Number MFCD00149548
InChI Key FPQYXAFKHLSWTI-UHFFFAOYSA-M
Synonym sodium pentane-1-sulfonate hydrate, 1-pentanesulfonic acid sodium salt monohydrate, sodium 1-pentanesulfonate monohydrate, potassium hydrate pentane-1-sulfonate, sodium 1-pentanesulfonate hydrate, sodium hydrate pentane-1-sulfonate, sodium pentane-1-sulfonate-water 1/1/1, 1-pentane sulphonic acid sodium salt monohydrate, 1-pentanesulfonic acid sodium salt monohydydrate, sodium 1-pentanesulfonate monohydrate, hplc grade
PubChem CID 23693099
IUPAC Name sodium;pentane-1-sulfonate;hydrate
SMILES CCCCCS(=O)(=O)[O-].O.[Na+]

Specifications

Absorbance (0.25M aq. soln.) (1 cm cell vs HPLC water), 0.05 max. at 220nm, 0.03 max. at 230nm, 0.02 max. at 240nm, 0.01 max. at 250nm, 0.01 max. at 260nm
Melting Point >300°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3(CH2)4SO3Na·H2O
Quantity 100 g
Beilstein 04, III, 23
Solubility Information Solubility in water: soluble
Formula Weight 192.22
Percent Purity 98+%
Physical Form Crystalline Solid
Chemical Name or Material 1-Pentanesulfonic acid, sodium salt monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • 1-Pentanesulfonic acid, sodium salt monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.