accessibility menu, dialog, popup

UserName

CAS RN 207683-22-5

BFFFFH2OH2ONa

CAS RN 207683-22-5

IUPAC Name: sodium tetrakis(4-fluorophenyl)boranuide dihydrate
Synonyms: sodium tetrakis(4-fluorophenyl)boranuide dihydrate
Molecular Weight (g/mol): 450.22
Molecular Formula: C24H20BF4NaO2
InChi Key: MSDGDEJOIBMWJD-UHFFFAOYSA-N
SMILES: O.O.[Na+].FC1=CC=C(C=C1)[B-](C1=CC=C(F)C=C1)(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results