accessibility menu, dialog, popup

UserName

CH3H3CBrOH

CAS RN 2077-19-2

IUPAC Name: 2-(4-bromophenyl)propan-2-ol
Synonyms: 2-(4-bromophenyl)propan-2-ol
Molecular Weight (g/mol): 215.09
Molecular Formula: C9H11BrO
InChi Key: AOGYBHJTXLXRSM-UHFFFAOYSA-N
SMILES: CC(C)(O)C1=CC=C(Br)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results