accessibility menu, dialog, popup

UserName

(R)S(S)(S)(S)(R)(S)(R)(R)OOOO(R)NHOH(R)HOHHHHNaMeMeMe

CAS RN 207737-97-1

IUPAC Name: sodium 2-(4-{7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)ethane-1-sulfonate hydrate
Synonyms: sodium 2-(4-{7,11-dihydroxy-9a,11a-dimethyl-tetradecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)ethanesulfonate hydrate
Molecular Weight (g/mol): 539.7
Molecular Formula: C26H46NNaO7S
InChi Key: OLPIZAYYAVQETM-UHFFFAOYNA-M
SMILES: O.[Na+].CC(CCC(=O)NCCS([O-])(=O)=O)C1CCC2C3CCC4CC(O)CCC4(C)C3CC(O)C12C
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)