missing translation for 'onlineSavingsMsg'
Learn More

Taurodeoxycholic acid, sodium salt hydrate, 97%

Product Code. 10716822 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10716822 1 g Glass bottle 1g
10011751 5 g Glass bottle 5g
10530034 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10716822 Supplier Thermo Scientific Acros Supplier No. 336840010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Taurodeoxycholic acid, sodium salt hydrate, 97%, CAS # 207737-97-1, also known as sodium taurodeoxylate, is a bile acid taurine conjugate of deoxycholic acid, and acts as a detergent. | CAS: 207737-97-1 | C26H44NNaO6S | 521.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Taurodeoxycholic acid, sodium salt hydrate, is a bile acid that is derived from deoxycholic acid

• It is also a conjugate acid of a taurodeoxycholate with detergent activity • It is formed in the liver by conjugating deoxycholate with a taurine group, commonly found in the form of a sodium salt

Applications

• Taurodeoxycholic acid, sodium salt hydrate has been studied to assess the adsorption of bile acids and salts in vegetable fibrous tissue

• It has been used as an anionic detergent important for protein solubilization

• This compound shows cholagogue and choleretic activities

• It has also been used to explore a methodology for determining conjugated and unconjugated bile salts both in serum and jejunal fluids

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207737-97-1
Molecular Formula C26H44NNaO6S
Molecular Weight (g/mol) 521.69
MDL Number MFCD00149238
InChI Key YXHRQQJFKOHLAP-FVCKGWAHSA-M
Synonym Sodium taurodeoxylate
PubChem CID 23702150
IUPAC Name sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate;hydrate
SMILES [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC(=O)NCCS([O-])(=O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
Specific Rotation +34.5° to +36.5° (20°C, 589nm) (c=2.5, water) (calculated on dry substance)
Water 7.0% max. (K.F.)
Formula Weight 521.69
Percent Purity ≥96.0%, 1.0% max. taurine, 1.0% max. sodium taurocholate, 1.0% max. sodium deocycholate (TLC)
Chemical Name or Material Taurodeoxycholic acid, sodium salt hydrate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Taurodeoxycholic acid, sodium salt hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.