accessibility menu, dialog, popup

UserName

FFO

CAS RN 208173-20-0

IUPAC Name: (2,3-difluorophenyl)(phenyl)methanone
Synonyms: (2,3-difluorophenyl)(phenyl)methanone
Molecular Weight (g/mol): 218.2
Molecular Formula: C13H8F2O
InChi Key: BWQOPMJTQPWHOZ-UHFFFAOYSA-N
SMILES: FC1=CC=CC(C(=O)C2=CC=CC=C2)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results