accessibility menu, dialog, popup

UserName

CAS RN 20825-89-2

OOOHO

CAS RN 20825-89-2

IUPAC Name: 3,4-dihydro-2H-1,5-benzodioxepine-7-carboxylic acid
Synonyms: 3,4-dihydro-2H-1,5-benzodioxepine-7-carboxylic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: MQSSBVLREFSMDP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=C(OCCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results