missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepine-7-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10304332
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10304332 1 g Amber Glass Bottle 1g
10252492 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10304332 Supplier Thermo Scientific Maybridge Supplier No. CC13201DA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 20825-89-2
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00276310
InChI Key MQSSBVLREFSMDP-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carboxylic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxylic acid, 2h-1,5-benzodioxepin-7-carboxylic acid, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-carboxylic acid, acmc-1crio, 7-carboxy-3,4-dihydro-2h-1,5-benzodioxepine, 2h-1,5-benzodioxepin-7-carboxylicacid, 3,4-dihydro, 2h-1,5-benzodioxepin-7-carboxylicacid,3,4-dihydro
PubChem CID 2776383
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepine-7-carboxylic acid
SMILES C1COC2=C(C=C(C=C2)C(=O)O)OC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-1, 5-benzodioxepine-7-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.