accessibility menu, dialog, popup

UserName

CH3OOOClN

CAS RN 208465-72-9

IUPAC Name: methyl 2-(chloromethyl)-1,3-oxazole-4-carboxylate
Synonyms: methyl 2-(chloromethyl)-1,3-oxazole-4-carboxylate
Molecular Weight (g/mol): 175.57
Molecular Formula: C6H6ClNO3
InChi Key: CMUKPCIZFMTLKD-UHFFFAOYSA-N
SMILES: COC(=O)C1=COC(CCl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results