accessibility menu, dialog, popup

UserName

H3CNONH2

CAS RN 20870-91-1

IUPAC Name: 5-amino-1-methyl-2,3-dihydro-1H-indol-2-one
Synonyms: 5-amino-1-methyl-3H-indol-2-one
Molecular Weight (g/mol): 162.19
Molecular Formula: C9H10N2O
InChi Key: ZGLUKQQSWABKDH-UHFFFAOYSA-N
SMILES: CN1C(=O)CC2=C1C=CC(N)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results