missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-methyl-2-oxoindoline, 97%, Thermo Scientific™

Product Code. 10295603
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10295603 250 mg Amber Glass Bottle 250mg
10295454 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10295603 Supplier Thermo Scientific Maybridge Supplier No. CC63014CB
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 20870-91-1
Molecular Formula C9H10N2O
Molecular Weight (g/mol) 162.19
MDL Number MFCD09702413
InChI Key ZGLUKQQSWABKDH-UHFFFAOYSA-N
Synonym 5-amino-1-methyl-2-oxoindoline, 5-amino-1-methylindolin-2-one, 5-amino-1-methyl-1,3-dihydro-2h-indol-2-one, 5-amino-1-methyl-2,3-dihydro-1h-indol-2-one, 2h-indol-2-one, 5-amino-1,3-dihydro-1-methyl, 5-azanyl-1-methyl-3h-indol-2-one, 5-amino-1-methyl-1,3-dihydro-indol-2-on, 5-amino-1-methyl-1,3-dihydro-indol-2-one, 2h-indol-2-one,5-amino-1,3-dihydro-1-methyl, 5-amino-1-methyl-1,3-dihydroindol-2-one
PubChem CID 22692470
IUPAC Name 5-amino-1-methyl-3H-indol-2-one
SMILES CN1C(=O)CC2=C1C=CC(N)=C2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 162.19
Percent Purity 90%
Chemical Name or Material 5-Amino-1-methyl-2-oxoindoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.