accessibility menu, dialog, popup

UserName

CAS RN 2095432-75-8

CH3ONNNH2HNOOHCl

CAS RN 2095432-75-8

IUPAC Name: N4-[(2H-1,3-benzodioxol-5-yl)methyl]-6-(3-methoxyphenyl)pyrimidine-2,4-diamine hydrochloride
Synonyms: N4-(2H-1,3-benzodioxol-5-ylmethyl)-6-(3-methoxyphenyl)pyrimidine-2,4-diamine hydrochloride
Molecular Weight (g/mol): 386.84
Molecular Formula: C19H19ClN4O3
InChi Key: XZOFNDFDGVAIEH-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=CC(=C1)C1=CC(NCC2=CC3=C(OCO3)C=C2)=NC(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results