missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AMBMP Hydrochloride, >98%

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Wnt canonical signaling activator. Also inhibits tubulin polymerization. Suppresses TLR2/4/5-induced inflammatory response in human monocytes. Inhibits cell proliferation and induces cell cycle arrest in MDA-MB-231 breast cancer cells. Also decreases embryo to blastocyst development.
Specifications
Specifications
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in DMSO |
| Formula Weight | 386.84 |
| Purity | >98% |
| Molecular Weight (g/mol) | 386.836 |
| Chemical Name or Material | N4-(1,3-Benzodioxol-5-ylmethyl)-6-(3-methoxyphenyl)-2,4-pyrimidinediamine hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction