accessibility menu, dialog, popup

UserName

CH3NONOOH3C

CAS RN 209912-44-7

IUPAC Name: methyl 4-(3-methyl-1,2,4-oxadiazol-5-yl)benzoate
Synonyms: methyl 4-(3-methyl-1,2,4-oxadiazol-5-yl)benzoate
Molecular Weight (g/mol): 218.21
Molecular Formula: C11H10N2O3
InChi Key: PJYMNVKGDLTDJE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C1=NC(C)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results