missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(3-methyl-1,2,4-oxadiazol-5-yl)benzoate, 97%, Thermo Scientific™

Product Code. 11340687
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11340687 250 mg Amber Glass Bottle 250mg
10506704 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340687 Supplier Thermo Scientific Maybridge Supplier No. CC50922CB
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 209912-44-7
Molecular Formula C11H10N2O3
Molecular Weight (g/mol) 218.212
MDL Number MFCD08690288
InChI Key PJYMNVKGDLTDJE-UHFFFAOYSA-N
Synonym methyl 4-3-methyl-1,2,4-oxadiazol-5-yl benzoate, 4-3-methyl-1,2,4-oxadiazole-5-yl benzoic acid methyl ester, benzoicacid, 4-3-methyl-1,2,4-oxadiazol-5-yl-, methyl ester
PubChem CID 11096067
IUPAC Name methyl 4-(3-methyl-1,2,4-oxadiazol-5-yl)benzoate
SMILES CC1=NOC(=N1)C2=CC=C(C=C2)C(=O)OC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 218.21
Percent Purity 97%
Chemical Name or Material Methyl 4-(3-methyl-1, 2, 4-oxadiazol-5-yl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.