accessibility menu, dialog, popup

UserName

CAS RN 211108-50-8

IUPAC Name: tert-butyl 3-fluoro-4-oxopiperidine-1-carboxylate
Synonyms: tert-butyl 3-fluoro-4-oxopiperidine-1-carboxylate
Molecular Weight (g/mol): 217.24
Molecular Formula: C10H16FNO3
InChi Key: JZNWQLLPLOQGOI-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCC(=O)C(F)C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results