missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 97%, Thermo Scientific Chemicals

Product Code. 13535010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13535010 1 g Glass bottle 1g
13525010 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13535010 Supplier Thermo Scientific Acros Supplier No. 446510010
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 211108-50-8
Molecular Formula C10H16FNO3
Molecular Weight (g/mol) 217.24
InChI Key JZNWQLLPLOQGOI-UHFFFAOYSA-N
Synonym 1-boc-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 1-boc-3-fluoro-4-piperidone, 1-tert-butoxycarbonyl-3-fluoro-4-piperidone, 1-piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxo-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl3-fluoro-4-oxopiperidine-1-carboxylate, tert-butyl 3-fluoro-4-oxo-piperidine-1-carboxylate, n-boc-3-fluoro-4-piperidinone
PubChem CID 10560711
IUPAC Name tert-butyl 3-fluoro-4-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=O)C(C1)F

Specifications

Melting Point 75°C to 77°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 217.24
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester
compliance-icons
Product Identifier
  • tert-Butyl 3-fluoro-4-oxo-1-piperidinecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: tert-Butyl 3-fluoro-4-oxo-1-piperidinecarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.