accessibility menu, dialog, popup

UserName

CH3FBr

CAS RN 212268-39-8

IUPAC Name: 1-(bromomethyl)-3-fluoro-5-methylbenzene
Synonyms: 1-(bromomethyl)-3-fluoro-5-methylbenzene
Molecular Weight (g/mol): 203.05
Molecular Formula: C8H8BrF
InChi Key: PVDKNKUVHSULSN-UHFFFAOYSA-N
SMILES: CC1=CC(CBr)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results