accessibility menu, dialog, popup

UserName

CAS RN 21248-45-3

OOSClCl

CAS RN 21248-45-3

IUPAC Name: 2-[(2,6-dichlorophenyl)sulfanyl]acetate
Synonyms: 2-[(2,6-dichlorophenyl)sulfanyl]acetate
Molecular Weight (g/mol): 236.09
Molecular Formula: C8H5Cl2O2S
InChi Key: URNRRSJGOOTQBO-UHFFFAOYSA-M
SMILES: [O-]C(=O)CSC1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results