accessibility menu, dialog, popup

UserName

CAS RN 2128-93-0

IUPAC Name: {[1,1'-biphenyl]-4-yl}(phenyl)methanone
Synonyms: {[1,1'-biphenyl]-4-yl}(phenyl)methanone
Molecular Weight (g/mol): 258.32
Molecular Formula: C19H14O
InChi Key: LYXOWKPVTCPORE-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results