accessibility menu, dialog, popup

UserName

CAS RN 21304-38-1

IH2NNH2

CAS RN 21304-38-1

IUPAC Name: 4-iodobenzene-1,2-diamine
Synonyms: 4-iodobenzene-1,2-diamine
Molecular Weight (g/mol): 234.04
Molecular Formula: C6H7IN2
InChi Key: FUOSRKZBOIVBOS-UHFFFAOYSA-N
SMILES: NC1=C(N)C=C(I)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results