accessibility menu, dialog, popup

UserName

CAS RN 21327-86-6

CH3ClHOO

CAS RN 21327-86-6

IUPAC Name: 2-chloro-6-methylbenzoic acid
Synonyms: 2-chloro-6-methylbenzoic acid
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: CEFMMQYDPGCYMG-UHFFFAOYSA-N
SMILES: CC1=CC=CC(Cl)=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results