accessibility menu, dialog, popup

UserName

BrCH3P

CAS RN 21406-61-1

IUPAC Name: pentyltriphenylphosphanium bromide
Synonyms: pentyltriphenylphosphanium bromide
Molecular Weight (g/mol): 413.34
Molecular Formula: C23H26BrP
InChi Key: VAUKWMSXUKODHR-UHFFFAOYSA-M
SMILES: [Br-].CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results