accessibility menu, dialog, popup

UserName

NNNBrF

CAS RN 214540-43-9

IUPAC Name: 1-benzyl-3-bromo-5-fluoro-1H-1,2,4-triazole
Synonyms: 1-benzyl-3-bromo-5-fluoro-1,2,4-triazole
Molecular Weight (g/mol): 256.08
Molecular Formula: C9H7BrFN3
InChi Key: MUYAKFNLCJMPDH-UHFFFAOYSA-N
SMILES: FC1=NC(Br)=NN1CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results