accessibility menu, dialog, popup

UserName

NH2N

CAS RN 214759-73-6

IUPAC Name: 1-[4-(piperidin-1-yl)phenyl]methanamine
Synonyms: 1-[4-(piperidin-1-yl)phenyl]methanamine
Molecular Weight (g/mol): 190.29
Molecular Formula: C12H18N2
InChi Key: WROBJEHXMLQDQP-UHFFFAOYSA-N
SMILES: NCC1=CC=C(C=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results