missing translation for 'onlineSavingsMsg'
Learn More

(4-Piperidinophenyl)methylamine, 97%, Thermo Scientific™

Product Code. 10286242
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10286242 1 g Amber Glass Bottle 1g
10194144 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10286242 Supplier Thermo Scientific Maybridge Supplier No. CC33913DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 214759-73-6
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD06659080
InChI Key WROBJEHXMLQDQP-UHFFFAOYSA-N
Synonym 4-piperidinophenyl methylamine, 4-piperidin-1-yl phenyl methanamine, 4-piperidinobenzylamine, 4-piperidin-1-yl-benzylamine, 1-4-piperidin-1-yl phenyl methanamine, 4-piperidin-1yl benzylamine, 4-piperidin-1ylphenyl methylamine, 4-piperidylphenyl methylamine, 4-1-piperidyl phenyl methanamine
PubChem CID 2795551
IUPAC Name (4-piperidin-1-ylphenyl)methanamine
SMILES NCC1=CC=C(C=C1)N1CCCCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 190.29
Percent Purity 97%
Chemical Name or Material (4-Piperidinophenyl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.