accessibility menu, dialog, popup

UserName

CH3OOOHHO

CAS RN 2150-43-8

IUPAC Name: methyl 3,4-dihydroxybenzoate
Synonyms: methyl 3,4-dihydroxybenzoate
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: CUFLZUDASVUNOE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(O)C(O)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)