accessibility menu, dialog, popup

UserName

CAS RN 21524-39-0

FNF

CAS RN 21524-39-0

IUPAC Name: 2,3-difluorobenzonitrile
Synonyms: 2,3-difluorobenzonitrile
Molecular Weight (g/mol): 139.11
Molecular Formula: C7H3F2N
InChi Key: GKPHNZYMLJPYJJ-UHFFFAOYSA-N
SMILES: FC1=CC=CC(C#N)=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)