accessibility menu, dialog, popup

UserName

NNH2BrCl

CAS RN 215364-85-5

IUPAC Name: 3-bromo-2-chloropyridin-4-amine
Synonyms: 3-bromo-2-chloropyridin-4-amine
Molecular Weight (g/mol): 207.46
Molecular Formula: C5H4BrClN2
InChi Key: KTWAUKYSQFZIET-UHFFFAOYSA-N
SMILES: NC1=C(Br)C(Cl)=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results