accessibility menu, dialog, popup

UserName

CAS RN 2157-56-4

H3CCH3NHONHO(E)(E)

CAS RN 2157-56-4

IUPAC Name: (E)-N-[(4E)-4-(hydroxyimino)pentan-2-ylidene]hydroxylamine
Synonyms: (E)-N-[(4E)-4-(hydroxyimino)pentan-2-ylidene]hydroxylamine
Molecular Weight (g/mol): 130.15
Molecular Formula: C5H10N2O2
InChi Key: WBRYLZHYOFBTPD-YDFGWWAZSA-N
SMILES: C\C(C\C(C)=N\O)=N/O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results