missing translation for 'onlineSavingsMsg'
Learn More

2,4-Pentanedione dioxime, 98+%, Thermo Scientific™

Product Code. 11439903
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439903 25 g 25g
11429903 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439903 Supplier Thermo Scientific Alfa Aesar Supplier No. A13003.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2157-56-4
Molecular Formula C5H10N2O2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00013932
InChI Key WBRYLZHYOFBTPD-YDFGWWAZSA-N
Synonym acetylacetone dioxime, 2,4-pentanedione dioxime, 2,4-pentanedione, dioxime, 2,4-pentanedioxime, 2,4-pentanedione oxime, 2,4-pentanedione,2,4-dioxime, 2,4-pentanedione, 2,4-dioxime, pentane-2,4-dione oxime, 2e,4e-2,4-pentanedione dioxime #, ne-n-4e-4-hydroxyiminopentan-2-ylidene hydroxylamine
PubChem CID 9573501
SMILES C\C(C\C(C)=N\O)=N/O

Specifications

Melting Point 147°C to 151°C
Quantity 25 g
Beilstein 4372703
IUPAC Name (NE)-N-[(4E)-4-hydroxyiminopentan-2-ylidene]hydroxylamine
Formula Weight 130.15
Percent Purity ≥98%
Chemical Name or Material 2,4-Pentanedione dioxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.