accessibility menu, dialog, popup

UserName

CAS RN 216144-09-1

IUPAC Name: 5-[3-(trifluoromethoxy)phenyl]-2H-1,2,3,4-tetrazole
Synonyms: 5-[3-(trifluoromethoxy)phenyl]-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 230.15
Molecular Formula: C8H5F3N4O
InChi Key: YKYWVXHVSVSQOX-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC(=C1)C1=NNN=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results