accessibility menu, dialog, popup

UserName

BH3CCH3OCH3HOOH

CAS RN 216393-63-4

IUPAC Name: [2-methoxy-5-(propan-2-yl)phenyl]boronic acid
Synonyms: 5-isopropyl-2-methoxyphenylboronic acid
Molecular Weight (g/mol): 194.04
Molecular Formula: C10H15BO3
InChi Key: UTKAEAGLVJFBMK-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=C1)C(C)C)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results