accessibility menu, dialog, popup

UserName

CAS RN 216393-67-8

IUPAC Name: 4-chloro-2-fluoro-6-iodoaniline
Synonyms: 4-chloro-2-fluoro-6-iodoaniline
Molecular Weight (g/mol): 271.46
Molecular Formula: C6H4ClFIN
InChi Key: SSNQXCONXNVTJN-UHFFFAOYSA-N
SMILES: NC1=C(I)C=C(Cl)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results