CAS RN 216755-57-6
CAS RN 216755-57-6
IUPAC Name:
1-bromo-3-(bromomethyl)-5-fluorobenzene
Synonyms:
1-bromo-3-(bromomethyl)-5-fluorobenzene
Molecular Weight (g/mol):
267.92
Molecular Formula:
C7H5Br2F
InChi Key:
DAUWIPUGOIFZNF-UHFFFAOYSA-N
SMILES:
FC1=CC(Br)=CC(CBr)=C1
1
–
1
of
1
results
1
3-Bromo-5-fluorobenzyl bromide, 95%, Thermo Scientific™
CAS: 216755-57-6 Molecular Formula: C7H5Br2F Molecular Weight (g/mol): 267.92 MDL Number: MFCD09258959 InChI Key: DAUWIPUGOIFZNF-UHFFFAOYSA-N Synonym: 3-bromo-5-fluorobenzyl bromide,1-bromo-3-bromomethyl-5-fluorobenzene,3-fluoro-5-bromobenzyl bromide,5-bromo-3-fluorobenzyl bromide,3-fluoro-5-bromobenzylbromide,5-bromo-3-bromomethyl-1-fluorobenzene,pubchem4886,acmc-20e3gr,3-bromo-5-fluorobenzylbromide,benzene,1-bromo-3-bromomethyl-5-fluoro PubChem CID: 23509619 IUPAC Name: 1-bromo-3-(bromomethyl)-5-fluorobenzene SMILES: FC1=CC(Br)=CC(CBr)=C1