accessibility menu, dialog, popup

UserName

H3CSOCH3

CAS RN 2168-93-6

IUPAC Name: 1-(butane-1-sulfinyl)butane
Synonyms: tetramethylene sulfoxide
Molecular Weight (g/mol): 162.29
Molecular Formula: C8H18OS
InChi Key: LOWMYOWHQMKBTM-UHFFFAOYSA-N
SMILES: CCCCS(=O)CCCC
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)