missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl sulfoxide, 93%, tech., Thermo Scientific™

Product Code. 10635332 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10635332 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10635332 Supplier Thermo Scientific Acros Supplier No. 108040250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2168-93-6
CAS Min % 91.0
CAS Max % 100.0
Melting Point 30.0°C to 35.0°C
Color Colorless
Density 0.8300g/mL
Boiling Point 250.0°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 93%
Packaging Glass bottle
Molecular Formula C8H18OS
Linear Formula [CH3(CH2)3]2SO
Refractive Index 1.4669
MDL Number MFCD00002093
Quantity 25 mL
Beilstein 01, 371
Specific Gravity 0.83
Synonym dibutyl sulfoxide, butyl sulfoxide, n-butyl sulfoxide, dibutyl sulphoxide, di-n-butyl sulfoxide, butane, 1,1'-sulfinylbis, sulfoxide, dibutyl, di-n-butylsulfoxide, butane,1-butylsulfinyl, butylsulfinyl butane
InChI Key LOWMYOWHQMKBTM-UHFFFAOYSA-N
SMILES CCCCS(=O)CCCC
IUPAC Name 1-butylsulfinylbutane
Molecular Weight (g/mol) 162.29
PubChem CID 16568
Formula Weight 162.29
Percent Purity 93%
Grade Technical
Physical Form Crystalline Solid
Chemical Name or Material n-Butyl sulfoxide, tech., 93%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.