accessibility menu, dialog, popup

UserName

CAS RN 2170-06-1

IUPAC Name: trimethyl(2-phenylethynyl)silane
Synonyms: trimethyl(2-phenylethynyl)silane
Molecular Weight (g/mol): 174.32
Molecular Formula: C11H14Si
InChi Key: UZIXCCMXZQWTPB-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results