missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-trimethylsilylacetylene, 99%

Product Code. 15462248
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462248 100 g 100g
15442248 5 g 5g
15452248 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15462248 Supplier Thermo Scientific Alfa Aesar Supplier No. H55866.22
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2170-06-1 | C11H14Si | 174.32 g/mol

1-Phenyl-2-trimethylsilylacetylene was used in the preparation of donor-stabilized Pt-η-alkyne complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-2-trimethylsilylacetylene was used in the preparation of donor-stabilized Pt-η-alkyne complexes.

Solubility
Sparingly Soluble (6.1E-3 g/L) (25°C)

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2170-06-1
Molecular Formula C11H14Si
Molecular Weight (g/mol) 174.32
MDL Number MFCD00054860
InChI Key UZIXCCMXZQWTPB-UHFFFAOYSA-N
SMILES C[Si](C)(C)C#CC1=CC=CC=C1

Specifications

Color Colorless
Density 0.886 g/mL
Boiling Point 87°C to 88°C (9 mmHg)
Flash Point 74°C (165°F)
Linear Formula C6H5C≡CSi(CH3)3
Refractive Index 1.528
Quantity 100 g
Beilstein 3088678
Synonym 1-phenyl-2-trimethylsilyl acetylene, trimethyl phenylethynyl silane, silane, trimethyl phenylethynyl, 1-phenyl-2-trimethylsilylacetylene, phenylethynyltrimethylsilane, trimethylsilyl phenylacetylene, trimethyl 2-phenylethynyl silane, phenylethynyl-trimethylsilane, phenylethynyl trimethylsilane, acmc-1bika
Solubility Information Sparingly Soluble (6.1E-3 g/L) (25°C)
IUPAC Name trimethyl(2-phenylethynyl)silane
PubChem CID 137464
Formula Weight 174.32
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-2-trimethylsilylacetylene
Show More Show Less

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.