accessibility menu, dialog, popup

UserName

CH3OOBrBrHO

CAS RN 21702-79-4

IUPAC Name: methyl 3,5-dibromo-2-hydroxybenzoate
Synonyms: methyl 3,5-dibromo-2-hydroxybenzoate
Molecular Weight (g/mol): 309.94
Molecular Formula: C8H6Br2O3
InChi Key: KISQISIKTRRMOX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=CC(Br)=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results