accessibility menu, dialog, popup

UserName

CAS RN 2174-64-3

H3COOHHO

CAS RN 2174-64-3

IUPAC Name: 5-methoxybenzene-1,3-diol
Synonyms: 3,5-dihydroxyanisole
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: HDVRLUFGYQYLFJ-UHFFFAOYSA-N
SMILES: COC1=CC(O)=CC(O)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)